
4-(Trifluoromethyl)nicotinic acid ≥98.0% (by GC, titration analysis)
: TCI
:158063-66-2
:
:C₇H₄F₃NO₂
:MFCD00082626
:191.11 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:137.0 to 141.0 deg-C
Frequently Bought Together