Tris[1-phenylisoquinoline-C2,N]iridium(III) ≥99.0% (by HPLC), purified by sublimation
Catalog # TCIAT2685-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to Amber to Dark red powder to crystal
- Purity (HPLC):min. 99.0 area%
Specification Test Results
About this item
:
:804.97 g/mol
CAS Number:435293-93-9
Formula:C₄₅H₃₀IrN₃
MDL Number:MFCD09842770