
Trimethyl[3-(triethoxysilyl)propyl]ammonium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:84901-27-9
:
Formula:C₁₂H₃₀ClNO₃Si
:299.91 g/mol
EINECS:284-463-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together