Order Entry
Denmark
ContactUsLinkComponent
Trimethylsulphonium hydroxide 0.2 mol/L in methanol methylating reagent
Flammable
Corrosive
Irritant
Health
Trimethylsulphonium hydroxide 0.2 mol/L in methanol methylating reagent
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Trimethylsulphonium hydroxide 0.2 mol/L in methanol methylating reagent
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Nonaqueous Titration):
    0.20 to 0.23 mol/L
  • High boiling impurities:
    to pass test

Specification Test Results

Related Information

Frequently Bought Together