Thioflavin T ≥95.0% (by HPLC)
TCIAT0558-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Lambda max.:410.0 to 414.0 nm (H2O)
- Absorbance (E1%1cm):min. 750 (H2O, 410.0 to 414.0 nm)
:
:318.86 g/mol
:219-228-9
:2390-54-7
:C₁₇H₁₉ClN₂S
:MFCD00011944