
Nickel(II) p-toluenesulphonate hexahydrate ≥98.0% (by titrimetric analysis)
: TCI
:6944-05-4
:
:C₇H₈O₃S
:MFCD00070542
:401.07 g/mol
:GHS08
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 % (calcd.on anh.substance)
- Water:20.0 to 22.0 %
Frequently Bought Together