DL-Thyronine ≥98.0% (by titrimetric analysis)
TCIAT0241-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
:
:273.29 g/mol
:213-854-6
:1034-10-2
:C₁₅H₁₅NO₄
:MFCD00045864