
2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3010-96-6
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00001329
:144.21 g/mol
EINECS:221-140-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 % (total of isomers)
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together