
Tetrabromo-o-sulphobenzoic anhydride ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:68460-01-5
:
Formula:C₇Br₄O₄S
MDL Number:MFCD00009745
:499.75 g/mol
EINECS:270-624-8
UN:1759
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Methanolysis Method):min. 97.0 %
- Melting point:216.0 to 224.0 deg-C
Specification Test Results
Frequently Bought Together