D-Biotin-N-succinimidyl ester ≥97.0% (by HPLC, total nitrogen)
: TCI
:35013-72-0
:
:C₁₄H₁₉N₃O₅S
:MFCD00078531
:341.38 g/mol
:140°C
:Freezer
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Protein labeling test (qualitative methoed):to pass test
Frequently Bought Together