Order Entry
Denmark
ContactUsLinkComponent
Sodium methoxide ∼5 mol/L in methanol
Flammable
Corrosive
Irritant
Health
Sodium methoxide ∼5 mol/L in methanol
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Sodium methoxide ∼5 mol/L in methanol
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    27.0 to 29.7 w/w%

Specification Test Results

Related Information

Frequently Bought Together