
Scopolamine hydrobromide trihydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6533-68-2
:
Formula:C₁₇H₂₁NO₄
MDL Number:MFCD00150066
:384.27 g/mol
Merck Index:14,08406
EINECS:204-050-6
UN:1544
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (after drying)
- Specific rotation [a]20/D:-24.0 to -27.0 deg (C=5, H2O) (after drying)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together