rSRL-Biotin for GlcNAc beta(1-2)Man, Gal beta(1-3)GalNAc
Catalog # TCIAR0233-1ML
Supplier: TCI
CAS Number:
Specifications
- Pk:1 mL
- Seq:2070
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Lowry Methoed):0.9 to 1.5 mg/mL
- Molecular weight:10 to 25 kDa
- Suitability for use in ELISA tests:min. 1.0 (10 micro g/mL, OD450)
Specification Test Results
About this item
:
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)