rPSL1a-Biotin for Sia alpha(2-6)Gal
TCIAR0230-1ML
: TCI
:
- Pk:1 mL
- Seq:2070
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Lowry Methoed):0.9 to 1.5 mg/mL
- Suitability for use in ELISA tests:min. 1.0 (1.0 micro g/mL, OD450)
- Molecular weight:25 to 40 kDa
:
:Ultra-Low Temperature Freezer (–80°C)