
Roxatidine Acetate hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:93793-83-0
:
Formula:C₁₉H₂₈N₂O₄·HCl
MDL Number:MFCD00941429
:384.90 g/mol
Merck Index:14,08274
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:146.0 to 150.0 deg-C
Specification Test Results
Frequently Bought Together