Rufinamide ≥98.0% (by HPLC)
: TCI
: 1-(2,6-Difluorobenzyl)-1H-1,2,3-triazole-4-carboxamide
:106308-44-5
:
:C₁₀H₈F₂N₄O
:MFCD00865314
:238.20 g/mol
:14,08293
:GHS07
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):22.50 to 24.50 %
- Melting point:239.0 to 243.0 deg-C
Frequently Bought Together