Order Entry
Denmark
ContactUsLinkComponent
Polyethylene glycol monododecyl ether n=∼23
Polyethylene glycol monododecyl ether n=∼23
Supplier:  TCI
Certificates
Polyethylene glycol monododecyl ether n=∼23
Supplier:  TCI
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White to Almost white powder to lump
  • Freezing point:
    33.0 to 38.0 deg-C
  • The average value of n:
    21.0 to 25.0