


Pyrogallol red solution for protein determination, ready-to-use
Catalog # TCIAP2575-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark red clear liquid
- Protein determination test:to pass test
Specification Test Results
About this item
:
:400.36 g/mol
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
:GHS02
:GHS07
:GHS08
CAS Number:32638-88-3
Formula:C₁₉H₁₂O₈S
MDL Number:MFCD00005046