Order Entry
Denmark
ContactUsLinkComponent
 
9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole ≥98.0% (by HPLC)
 :  TCI
 
undefined
9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole ≥98.0% (by HPLC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %

 

Frequently Bought Together