Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

γ-(N-Morpholino)propanesulphonic acid (MOPS)

Marketsource
Supplier:   Serva
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Supplier:  Serva
Marketsource
Certificates
γ-(N-Morpholino)propanesulphonic acid (MOPS)

3 Options Available Below

Product Details & Documents
Synonyms:  3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
  • CAS Number:1132-61-2
  • Formula:C₇H₁₅NO₄S
  • MDL Number:MFCD00006183
  • Molecular Weight:209.27 g/mol
  • Merck Index:13,06288
  • Melting Pt:> 250 °C
  • Flash Pt:110 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:214-478-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 328,58 kr
    328,58 kr
    1 * 100 g
       328,58 kr

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1.060,40 kr
    1.060,40 kr
    1 * 500 g
       1.060,40 kr

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1.874,38 kr
    1.874,38 kr
    1 * 1 kg
       1.874,38 kr

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.