Order Entry
Denmark
ContactUsLinkComponent
 
Pyritinol ≥96.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
Pyritinol ≥96.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (Nonaqueous Titration):
    min. 96.0 %

 

Frequently Bought Together