Order Entry
Denmark
ContactUsLinkComponent
Polyethylene glycol 2,000
Polyethylene glycol 2,000
Supplier:  TCI
Certificates
Polyethylene glycol 2,000
Supplier:  TCI
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Melting point:
    51.0 to 55.0 deg-C
  • Average molecular weight:
    1,850 to 2,150

Specification Test Results

Frequently Bought Together