Pyrogallol red ≥95.0% (by HPLC) for protein research
Catalog # TCIAP1976-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Dark red to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Lambda max.:539.0 to 543.0 nm (pH 8.0 buffer sol.)
- Absorbance (E1%1cm):min. 850 (pH 8.0 buffer sol., 539.0 to 543.0 nm)
- Suitability for protein-analysis:to pass test
Specification Test Results
About this item
:
:400.36 g/mol
EINECS:251-134-3
CAS Number:32638-88-3
Formula:C₁₉H₁₂O₈S
MDL Number:MFCD00005046