Order Entry
Denmark
ContactUsLinkComponent
Polyethyleneimine ∼30% in water for biochemical research
Corrosive
Polyethyleneimine ∼30% in water for biochemical research
Supplier:  TCI
Certificates
Corrosive
Polyethyleneimine ∼30% in water for biochemical research
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Total Nitrogen:
    9.2 to 10.5 %
  • Nucleic acid precipitation test:
    to pass test

Specification Test Results

Related Information

Frequently Bought Together