4-Propylcyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:52204-65-6
:
Formula:C₉H₁₈O
MDL Number:MFCD00091618
:142.24 g/mol
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
Specification Test Results
Frequently Bought Together