Pyrrolidin-2-ylmethanol ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:498-63-5
:
Formula:C5H11NO
MDL Number:MFCD00601073
:101.15 g/mol
Boiling Pt:76°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together