Order Entry
Denmark
ContactUsLinkComponent
Palladium 5% on carbon wetted with ca. 55% water
Palladium 5% on carbon wetted with ca. 55% water
Supplier:  TCI
Certificates
Palladium 5% on carbon wetted with ca. 55% water
Supplier:  TCI

Specification Test Results

  • Appearance:
    Black powder to crystal
  • Palladium:
    4.9 to 5.1 %

Specification Test Results

Frequently Bought Together