4-Phenylethynylphthalic anhydride ≥98.0% (by GC, titration analysis)
: TCI
:119389-05-8
:MFCD10574892
:C₁₆H₈O₃
:MFCD01862632
:248.24 g/mol
:GHS07
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Morpholine Method):min. 98.0 %
- Melting point:149.0 to 154.0 deg-C
Frequently Bought Together