Polyethylene glycol mono-4-octylphenyl ether n=∼10
Supplier: TCI
Certificates
About this item
Synonyms: Triton X-100, Polyethylene glycol mono(4-tert-octylphenyl) ether
CAS Number:9002-93-1
:
Formula:C₈H₁₇C₆H₄(OCH₂CH₂)nOH
MDL Number:MFCD00132505
Merck Index:14,06761
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- The average value of n:8.5 to 11.5
Specification Test Results
Frequently Bought Together