Order Entry
Denmark
ContactUsLinkComponent
Polyethylene glycol mono-4-nonylphenyl ether n=∼15
Irritant
Polyethylene glycol mono-4-nonylphenyl ether n=∼15
Katalog # TCIAP0708-25G
Supplier:  TCI
CAS Number:  
Irritant
Polyethylene glycol mono-4-nonylphenyl ether n=∼15
Katalog # TCIAP0708-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Acid Value:
    max. 1.0
  • The average value of n:
    13.0 to 17.0

Specification Test Results

About this item

 :
Merck Index:14,06677
EINECS:500-045-0
 :GHS07
CAS Number:26027-38-3
MDL Number:MFCD00148006