
Polyethylene glycol mono-4-nonylphenyl ether n=∼2
Catalog # TCIAP0704-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- The average value of n:1.8 to 2.5
Specification Test Results
About this item
:
Merck Index:14,06677
EINECS:243-816-4
:GHS07
CAS Number:26027-38-3
MDL Number:MFCD00148006