Order Entry
Denmark
ContactUsLinkComponent
β-Methylstyrene (cis- and trans- mixture) cis- and trans-mixture ≥95.0% (by GC) stabilised
Flammable
β-Methylstyrene (cis- and trans- mixture) cis- and trans-mixture ≥95.0% (by GC) stabilised
Catalog # TCIAP0495-25ML
Supplier:  TCI
CAS Number:  
Flammable
β-Methylstyrene (cis- and trans- mixture) cis- and trans-mixture ≥95.0% (by GC) stabilised
Catalog # TCIAP0495-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomer)

Specification Test Results

About this item

 :
 :118.18 g/mol
Boiling Pt:177°C
Flash Pt:60°C
ADR:3,III
 :Stabilised with 4-tert-butylpyrocatechol (TBC)
EINECS:211-287-9
UN:3295
 :GHS02
CAS Number:637-50-3
Formula:C₉H₁₀
MDL Number:MFCD00009280