Order Entry
Denmark
ContactUsLinkComponent
p-Phenetidine hydrochloride ≥98.0% (by HPLC, titration analysis)
p-Phenetidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP0116-25G
Supplier:  TCI
CAS Number:  
p-Phenetidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP0116-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Solubility in Water:
    very faint turbidity

Specification Test Results

About this item

 :
 :173.64 g/mol
EINECS:211-292-6
CAS Number:637-56-9
Formula:C₈H₁₁NO
MDL Number:MFCD00035503