Sodium palmitate ≥97.0% (by GC, titration analysis)
: TCI
:408-35-5
:
:C₁₆H₃₂O₂
:MFCD00002749
:278.41 g/mol
:206-988-1
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Solubility in hot EtOH (50vol%):very faint turbidity
Frequently Bought Together