N-Acetylneuraminic acid tetramer alpha(2-8) ≥95.0% (by HPLC)
TCIAN1165-5MG
: TCI
:
- Pk:5 mg
- Seq:2210
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
:
:1183.04 g/mol
:Freezer
:96425-83-1
:C₄₄H₇₀N₄O₃₃