N-Acetylneuraminic acid trimer alpha(2-8) ≥95.0% (by HPLC)
Catalog # TCIAN1164-10MG
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
Specification Test Results
About this item
:
:891.78 g/mol
Storage Temperature:Freezer
CAS Number:76152-09-5
Formula:C₃₃H₅₃N₃O₂₅