Order Entry
Denmark
ContactUsLinkComponent
 
N-Acetylneuraminic acid Dimer alpha(2-8) ≥95.0% (by HPLC)
  TCIAN1163-10MG
 :  TCI
 :  
N-Acetylneuraminic acid Dimer alpha(2-8) ≥95.0% (by HPLC)
  TCIAN1163-10MG
 :  TCI
 :  

 

  • Pk:
    10 mg
  • Seq:
    2275

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

 

 

 :
 :600.53 g/mol
 :Freezer
 :95983-78-1
 :C₂₂H₃₆N₂O₁₇