Order Entry
Denmark
ContactUsLinkComponent
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Irritant
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together