
1H,1H,2H,2H-Nonafluorohexyl methacrylate ≥98.0% (by GC) stabilised
: TCI
:1799-84-4
:
:C₁₀H₉F₉O₂
:MFCD00236094
:62°C
:79°C
:Stabilised with 4-methoxyphenol (MEHQ)
:332.17 g/mol
:217-287-5
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together