Order Entry
Denmark
ContactUsLinkComponent
4-Nitrophenyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC) substrate for alpha-Mannosidase
4-Nitrophenyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC) substrate for alpha-Mannosidase
Catalog # TCIAN0619-1G
Supplier:  TCI
CAS Number:  
4-Nitrophenyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC) substrate for alpha-Mannosidase
Catalog # TCIAN0619-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    178.0 to 183.0 deg-C
  • Specific rotation [a]20/D:
    +154 to +166 deg (C=0.2, H2O)

Specification Test Results

About this item

 :
 :301.25 g/mol
EINECS:233-776-6
CAS Number:10357-27-4
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00066002