Order Entry
Denmark
ContactUsLinkComponent
 
 
(S)-(-)-N-(5-Nitro-2-pyridyl)prolinol ≥99.0% (by titrimetric analysis)
  TCIAN0540-1G
 :  TCI
 :  
 
(S)-(-)-N-(5-Nitro-2-pyridyl)prolinol ≥99.0% (by titrimetric analysis)
  TCIAN0540-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Melting point:
    81.0 to 84.0 deg-C
  • Specific rotation [a]20/D:
    -161.0 to -166.0 deg (C=1, EtOH)

 

 

 :
 :223.23 g/mol
 :GHS07
 :88374-37-2
 :C₁₀H₁₃N₃O₃
 :MFCD00145250