N-Methoxycarbonyl-D-phenylglycine ≥97.0% (by HPLC, titration analysis)
: TCI
:50890-96-5
:
:C₁₀H₁₁NO₄
:MFCD12796004
:209.20 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:117.0 to 121.0 deg-C
- Specific rotation [a]20/D:+3.0 to +5.0 deg (C=1, H2O)
Frequently Bought Together