1-Methyl-1-n-octylpyrrolidinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:927021-43-0
:
Formula:C₁₅H₂₈F₆N₂O₄S₂
MDL Number:MFCD09038851
:478.51 g/mol
Melting Pt:–12°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
Frequently Bought Together