Order Entry
Denmark
ContactUsLinkComponent
 
Irritant
Health
N-Methyl-2-pyrrolidone (NMP), low water content ≥99.0% (by GC)
Catalog # TCIAM3055-500ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
N-Methyl-2-pyrrolidone (NMP), low water content ≥99.0% (by GC)
Catalog # TCIAM3055-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Water:
    max. 300 ppm

Specification Test Results

About this item

 :
 :99.13 g/mol
Merck Index:14,06117
Boiling Pt:202°C
Melting Pt:-24°C
Melting Pt:–24°C
Flash Pt:95°C
EINECS:212-828-1
 :GHS07
 :GHS08
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193