
4-Methyl-2-phenylpyridine ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3475-21-6
:
Formula:C₁₂H₁₁N
MDL Number:MFCD00087695
:169.23 g/mol
Boiling Pt:115°C
EINECS:222-448-8
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:47.0 to 51.0 deg-C
- melting range:max. 5.0 deg-C
Specification Test Results
Frequently Bought Together