
2-Methylene-1,3-propanediol ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3513-81-3
:
Formula:C₄H₈O₂
MDL Number:MFCD00075162
:88.11 g/mol
EINECS:222-516-7
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together