Order Entry
Denmark
ContactUsLinkComponent
2-Methoxy-5-methylphenylboronic acid (contains varying amounts of Anhydride)
Irritant
2-Methoxy-5-methylphenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
undefined
Irritant
2-Methoxy-5-methylphenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    97.0 to 113.0 %

Specification Test Results

Related Information

Frequently Bought Together