Order Entry
Denmark
ContactUsLinkComponent
4-(4-Methylpiperazin-1-yl)benzoic acid ≥98.0% (by HPLC, titration analysis)
4-(4-Methylpiperazin-1-yl)benzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
4-(4-Methylpiperazin-1-yl)benzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together