Methylmagnesium bromide ∼30% ∼3 mol/L in MeTHF (2-methyltetrahydrofuran)
Catalog # TCIAM2237-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):30.0 to 34.5 w/w%
Specification Test Results
About this item
:
:119.24 g/mol
EINECS:200-844-1
UN:3399
ADR:4.3,I
:GHS02
:GHS07
CAS Number:75-16-1
Formula:CH₃BrMg
MDL Number:MFCD00000041