Order Entry
Denmark
ContactUsLinkComponent
2-(4-Methoxyphenyl)pyridine ≥97.0% (by GC, titration analysis)
Irritant
2-(4-Methoxyphenyl)pyridine ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
2-(4-Methoxyphenyl)pyridine ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    53.0 to 57.0 deg-C

Specification Test Results

Frequently Bought Together